Learn More
Medchemexpress LLC Akn-028 (acetate) | 98.38% | 362.39 | 100 MG
SDP
Catalog No. 5000436283
Encompass_Preferred
AKN-028 acetate is a novel tyrosine kinase (TK) inhibitor. It is a potent, orally active FMS-like receptor tyrosine kinase 3 (FLT3) inhibitor with an IC50 value of 6 nM. It inhibits FLT3 autophosphorylation and induces dose-dependent cytotoxic response (mean IC50=1 μM). AKN-028 acetate induces apoptosis by activation of caspase 3 and can be used in research of acute myeloid leukemia (AML).
  • Potent and orally active FLT3 inhibitor with an IC50 of 6 nM.
  • Inhibits FLT3 autophosphorylation.
  • Induces dose-dependent cytotoxic response with a mean IC50 of 1 μM.
  • Induces apoptosis through activation of caspase 3.
  • Suitable for research in acute myeloid leukemia (AML).
  • Shows cytotoxic activity against AML cell lines.
  • Inhibits tumor growth in xenograft models.
Catalog No. 50-004-36283 Supplier Medchemexpress LLC Supplier No. HY118304B100MG
AKN-028 acetate is a novel tyrosine kinase (TK) inhibitor. It is a potent, orally active FMS-like receptor tyrosine kinase 3 (FLT3) inhibitor with an IC50 value of 6 nM. It inhibits FLT3 autophosphorylation and induces dose-dependent cytotoxic response (mean IC50=1 μM). AKN-028 acetate induces apoptosis by activation of caspase 3 and can be used in research of acute myeloid leukemia (AML).
  • Potent and orally active FLT3 inhibitor with an IC50 of 6 nM.
  • Inhibits FLT3 autophosphorylation.
  • Induces dose-dependent cytotoxic response with a mean IC50 of 1 μM.
  • Induces apoptosis through activation of caspase 3.
  • Suitable for research in acute myeloid leukemia (AML).
  • Shows cytotoxic activity against AML cell lines.
  • Inhibits tumor growth in xenograft models.