Learn More
Cayman Chemical AKOS B018304, Purity: ≥98%, CAS: 6308-22-1, 10 mg
SDP
Catalog No. 502910803
Encompass_Preferred
A derivative of 1,3-thiazolidin-4-one; inhibits the binding of ΔFosB to an oligonucleotide containing the Cdk5-AP-1 consensus site in a cell-free assay (IC50 = 10.1 µM); induces ΔFosB- and ΔFosB/JunD-mediated gene transcription in a reporter assay using Neuro2a cells at 12.5 µM
Catalog No. 50-291-0803 Supplier Cayman Chemical Supplier No. 3569410 MG
A derivative of 1,3-thiazolidin-4-one; inhibits the binding of ΔFosB to an oligonucleotide containing the Cdk5-AP-1 consensus site in a cell-free assay (IC50 = 10.1 µM); induces ΔFosB- and ΔFosB/JunD-mediated gene transcription in a reporter assay using Neuro2a cells at 12.5 µM