Learn More
Aviva Systems Biology Corp AKR1A1 Antibody - C-terminal region (ARP64535_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64535_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKR1A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human AKR1A1. Alternate names: ALDR1, ALR, ARM, DD3, MGC12529, MGC1380ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-289