Learn More
Aviva Systems Biology Corp AKR1C1 Antibody - N-terminal region (ARP60165_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60165_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKR1C1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AKR1C1. Alternate names: 2-ALPHA-HSD, 20-ALPHA-HSD, C9, DD1, DDH, DDH1, H-37, HAKRC, MBAB, MGC8954, HBAB, DD1/DD2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-967