Learn More
Aviva Systems Biology Corp AKR1C2 Antibody - N-terminal region (ARP53507_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53507_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKR1C2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AKR1C2. Alternate names: AKR1C-pseudo, BABP, DD, DD2, DDH2, HAKRD, HBAB, MCDR2, SRXY8ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-63037