Learn More
MyBioSource Aldo-Keto Reductase Family 1 Member C3, Active Protein
SDP
Supplier:  MyBioSource MBS1435331MG
Encompass
The product is human Aldo-Keto Reductase Family 1 Member C3 (AKR1C3) protein expressed in human cells with a molecular weight of 37kDa. It is supplied as a lyophilized powder and is ≥95% pure as determined by SDS-PAGE. The protein can be used for a variety of biochemical and cell-based assays to study its enzymatic activity and biological function. Recommended storage is at -20°C. This product would be useful for researchers studying human metabolism, steroidogenesis, and cancer biology.
Catalog No. NC2746570