Learn More
Aviva Systems Biology Corp AKR7A2 Antibody - N-terminal region (ARP65054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKR7A2. It was validated on Western Blot. . Alternate names: AFAR, AFAR1, AFB1-AR1, AKR7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-417