Learn More
Aviva Systems Biology Corp AKR7A3 Antibody - middle region (ARP54824_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54824_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKR7A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AKR7A3. Alternate names: AFAR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61330