Learn More
MSC Akro-Mils Bin Divider, Long, Polypropylene, Black, 6 Pack, for Akro-Grid 33228
Catalog No. 5000536484
Encompass
Akro-Mils bin dividers are designed for use with Akro-Grid dividable containers, specifically the 33228 model, allowing for the creation of custom-sized storage compartments. These long dividers are made from durable polypropylene and are ideal for work-in-process transfer applications. Each package includes a convenient 6-pack of black dividers.
  • Allows subdivision of storage containers up to a 1-1/8 inch square.
  • Creates custom sized storage compartments.
  • Ideal for work-in-process transfer applications.
  • Made from polypropylene for durability.
  • Comes in a pack of 6.
  • Black color.
  • Features divider slots for numerous compartment size configurations.
Catalog No. 50-005-36484 Supplier MSC Supplier No. 89805535
Akro-Mils bin dividers are designed for use with Akro-Grid dividable containers, specifically the 33228 model, allowing for the creation of custom-sized storage compartments. These long dividers are made from durable polypropylene and are ideal for work-in-process transfer applications. Each package includes a convenient 6-pack of black dividers.
  • Allows subdivision of storage containers up to a 1-1/8 inch square.
  • Creates custom sized storage compartments.
  • Ideal for work-in-process transfer applications.
  • Made from polypropylene for durability.
  • Comes in a pack of 6.
  • Black color.
  • Features divider slots for numerous compartment size configurations.