Learn More
MSC Akro-Mils Bin Divider, Polypropylene, Black, 7.2 inch
Catalog No. 5000515525
Encompass
This bin divider is made from polypropylene and is designed for use with dividable containers. It is ideal for work-in-process transfer applications, allowing users to create custom-sized storage compartments as small as 1-1/8 "
  • Dividers can be used to create custom sized storage compartments
  • Divider slots allow for the creation of hundreds of different compartment sizes
  • Made from recycled content
Catalog No. 50-005-15525 Supplier MSC Supplier No. 89805527
This bin divider is made from polypropylene and is designed for use with dividable containers. It is ideal for work-in-process transfer applications, allowing users to create custom-sized storage compartments as small as 1-1/8 "
  • Dividers can be used to create custom sized storage compartments
  • Divider slots allow for the creation of hundreds of different compartment sizes
  • Made from recycled content