Learn More
Enterprise Technology Solutions Akro-Mils Bins, Unbreakable/Waterproof, 4-1/8"x5-3/8"x3", Blue
SDP
Catalog No. 5000832550
Encompass_Preferred
Develop an efficient storage system using these versatile bins that stack with each other or sit on shelving. Take back valuable floor space by hanging from Akro-Mils louvered panel (sold separately) hanging systems. Injection-molded, blue industrial-grade polymer bins are virtually unbreakable and waterproof.
Catalog No. 50-008-32550 Supplier Enterprise Technology Solutions Supplier No. AKM30210B
Develop an efficient storage system using these versatile bins that stack with each other or sit on shelving. Take back valuable floor space by hanging from Akro-Mils louvered panel (sold separately) hanging systems. Injection-molded, blue industrial-grade polymer bins are virtually unbreakable and waterproof.