Learn More
MSC Akro-Mils Dividable Plastic Storage Box, 10-7/8 x 16-1/2 x 6 in, Gray, Plastic
Catalog No. 5000528254
Encompass
Akro-Mils Dividable Plastic Storage Boxes are suitable for work-in-process transfer applications or for sorting inventory. Made from durable plastic, these gray rectangular bins feature strong stacking rims and external ribbing for added strength. They offer a 35 lb load capacity and resist most solvents and chemicals.
  • Ideal for work-in-process transfer applications or sorting inventory.
  • Dividers can create custom-sized storage compartments.
  • Strong stacking rims and external ribbing provide added strength.
  • Optional snap-on lids keep contents secure and dust-free.
  • Resists most solvents and chemicals.
  • Stacking design allows for efficient storage.
  • Fits on shelving for organized inventory.
  • Lids are available as accessories.
Catalog No. 50-005-28254 Supplier MSC Supplier No. 43395169
Akro-Mils Dividable Plastic Storage Boxes are suitable for work-in-process transfer applications or for sorting inventory. Made from durable plastic, these gray rectangular bins feature strong stacking rims and external ribbing for added strength. They offer a 35 lb load capacity and resist most solvents and chemicals.
  • Ideal for work-in-process transfer applications or sorting inventory.
  • Dividers can create custom-sized storage compartments.
  • Strong stacking rims and external ribbing provide added strength.
  • Optional snap-on lids keep contents secure and dust-free.
  • Resists most solvents and chemicals.
  • Stacking design allows for efficient storage.
  • Fits on shelving for organized inventory.
  • Lids are available as accessories.