Learn More
MSC Akro-Mils Hopper Shelf Bin, 4-1/8 in Wide, Plastic, Yellow
Catalog No. 5000528537
Encompass
This hopper shelf bin is designed for organized storage in various industrial and laboratory settings. Constructed from durable yellow plastic, it features a hopper front for easy access to contents. The bin's dimensions are 4-1/8 in wide x 11-5/8 in deep x 4 in high.
  • Nestable design
  • Built-in rear hang-lock
  • Durable construction
  • Fits on standard wire or steel shelving
  • Hopper front for easy access
  • Molded-in label area identifies content
  • Waterproof and resistant to grease or oil
  • Functions in temperatures down to -20 °F
Catalog No. 50-005-28537 Supplier MSC Supplier No. 43235878
This hopper shelf bin is designed for organized storage in various industrial and laboratory settings. Constructed from durable yellow plastic, it features a hopper front for easy access to contents. The bin's dimensions are 4-1/8 in wide x 11-5/8 in deep x 4 in high.
  • Nestable design
  • Built-in rear hang-lock
  • Durable construction
  • Fits on standard wire or steel shelving
  • Hopper front for easy access
  • Molded-in label area identifies content
  • Waterproof and resistant to grease or oil
  • Functions in temperatures down to -20 °F