Learn More
MSC Akro-Mils Plastic Hopper Shelf Bin, Red, 11-1/8 x 11-5/8 x 4 in
Catalog No. 5000527054
Encompass
This durable 4-inch-tall shelf bin is made from industrial-grade polymers, featuring a wide hopper front for easy access. These one-piece, waterproof bins are designed to outlast corrugated alternatives and are sized to fit on standard wire or steel shelving.
  • Reinforced side ribs for added strength.
  • Built-in rear hanglock allows bin to tilt out for complete access.
  • Bins nest when empty.
  • Capacity of 197 cubic inches.
Catalog No. 50-005-27054 Supplier MSC Supplier No. 43376045
This durable 4-inch-tall shelf bin is made from industrial-grade polymers, featuring a wide hopper front for easy access. These one-piece, waterproof bins are designed to outlast corrugated alternatives and are sized to fit on standard wire or steel shelving.
  • Reinforced side ribs for added strength.
  • Built-in rear hanglock allows bin to tilt out for complete access.
  • Bins nest when empty.
  • Capacity of 197 cubic inches.