Learn More
MSC Akro-Mils Plastic Hopper Shelf Bin: Yellow, Polymer, 11-1/8 x 23-5/8 x 4 in
Catalog No. 5000532567
Encompass
This durable 4-inch-tall shelf bin is made from industrial-grade polymers. Its wide hopper front allows for easy access. These one-piece, waterproof bins are designed to outlast corrugated bins and are sized to fit on standard wire or steel shelving.
  • Reinforced side ribs for added strength
  • Built-in rear hang-lock allows the bin to tilt out for complete access
  • Bins nest when empty
  • Features a large label area aid
  • Hopper front provides easy access
  • Designed to fit on shelving
  • Durable construction
  • Load capacity of 20 lb
  • Inside capacity of 513 cu. in.
Catalog No. 50-005-32567 Supplier MSC Supplier No. 43376557
This durable 4-inch-tall shelf bin is made from industrial-grade polymers. Its wide hopper front allows for easy access. These one-piece, waterproof bins are designed to outlast corrugated bins and are sized to fit on standard wire or steel shelving.
  • Reinforced side ribs for added strength
  • Built-in rear hang-lock allows the bin to tilt out for complete access
  • Bins nest when empty
  • Features a large label area aid
  • Hopper front provides easy access
  • Designed to fit on shelving
  • Durable construction
  • Load capacity of 20 lb
  • Inside capacity of 513 cu. in.