Learn More
MSC Akro-Mils Plastic Storage Bin, 10.875 in L, 11 in W, 5 in H
Catalog No. 5000528662
Encompass
This storage bin is an industry-leading hanging and stacking solution. Heavy-duty, these bins stack together or hang from louvered racks and panels, rail racks, and louvered carts for high-density storage. They are corrosion-proof and unaffected by weak acids and alkalis. Length dividers and clear lids are available.
  • Heavy-duty construction
  • Hangs from louvered racks and panels, rail racks, and louvered carts for high-density storage
  • Stacks together for convenient storage
  • Corrosion-proof and unaffected by weak acids and alkalis
  • Fits on shelving
  • Features a hopper front design
  • Compatible with length dividers and clear lids (accessories sold separately)
Catalog No. 50-005-28662 Supplier MSC Supplier No. 43386150
This storage bin is an industry-leading hanging and stacking solution. Heavy-duty, these bins stack together or hang from louvered racks and panels, rail racks, and louvered carts for high-density storage. They are corrosion-proof and unaffected by weak acids and alkalis. Length dividers and clear lids are available.
  • Heavy-duty construction
  • Hangs from louvered racks and panels, rail racks, and louvered carts for high-density storage
  • Stacks together for convenient storage
  • Corrosion-proof and unaffected by weak acids and alkalis
  • Fits on shelving
  • Features a hopper front design
  • Compatible with length dividers and clear lids (accessories sold separately)