Learn More
MSC Akro-Mils Plastic Storage Bin, 5-3/8 in Length, 4-1/8 in Width, 10 lb Capacity
Catalog No. 5000528539
Encompass
These plastic storage bins are designed for hanging and stacking. They feature heavy-duty construction and can be used with louvered racks, panels, rail racks, and louvered carts for high-density storage. The bins are corrosion-proof and unaffected by weak acids and alkalis.
  • Heavy-duty construction for durability
  • Stackable design for efficient space utilization
  • Can be hung on racks and panels
  • Corrosion-proof and resistant to weak acids and alkalis
  • Hopper front provides easy access to contents
  • Compatible with shelving systems
  • Load capacity of 10 lb
Catalog No. 50-005-28539 Supplier MSC Supplier No. 43380054
These plastic storage bins are designed for hanging and stacking. They feature heavy-duty construction and can be used with louvered racks, panels, rail racks, and louvered carts for high-density storage. The bins are corrosion-proof and unaffected by weak acids and alkalis.
  • Heavy-duty construction for durability
  • Stackable design for efficient space utilization
  • Can be hung on racks and panels
  • Corrosion-proof and resistant to weak acids and alkalis
  • Hopper front provides easy access to contents
  • Compatible with shelving systems
  • Load capacity of 10 lb