Learn More
MSC Akro-Mils Shelf Bin, Polymer, Yellow, 4 in Height, Hopper Front
Catalog No. 5000533686
Encompass
These durable 4-inch-tall shelf bins are constructed from industrial-grade polymers, offering a neat and attractive storage solution. They feature a wide hopper front for easy access and are one-piece waterproof bins that outlast corrugated alternatives. Designed to fit on standard wire or steel shelving, they are unaffected by grease or oil and can function in temperatures down to -20°F.
  • Constructed from durable industrial-grade polymers.
  • Wide hopper front provides easy access.
  • One-piece waterproof construction outlasts corrugated bins.
  • Designed to fit standard wire or steel shelving.
  • Unaffected by grease or oil.
  • Functions in temperatures down to -20°F.
  • Nests when empty.
  • Reinforced side ribs for added strength.
  • Built-in rear hang-lock allows bin to tilt out for complete access.
Catalog No. 50-005-33686 Supplier MSC Supplier No. 43376805
These durable 4-inch-tall shelf bins are constructed from industrial-grade polymers, offering a neat and attractive storage solution. They feature a wide hopper front for easy access and are one-piece waterproof bins that outlast corrugated alternatives. Designed to fit on standard wire or steel shelving, they are unaffected by grease or oil and can function in temperatures down to -20°F.
  • Constructed from durable industrial-grade polymers.
  • Wide hopper front provides easy access.
  • One-piece waterproof construction outlasts corrugated bins.
  • Designed to fit standard wire or steel shelving.
  • Unaffected by grease or oil.
  • Functions in temperatures down to -20°F.
  • Nests when empty.
  • Reinforced side ribs for added strength.
  • Built-in rear hang-lock allows bin to tilt out for complete access.