Learn More
MSC Akro-Mils Shelf Bin, Polypropylene, Stone, 10-7/8 x 4-1/8 x 4 in.
Catalog No. 5000529514
Encompass
This shelf bin is constructed from durable polypropylene and designed for efficient storage. It features a stone color and outer dimensions of 10-7/8 in. L x 4-1/8 in. W x 4 in. H, making it suitable for various organizational needs.
  • Can be hung or stacked for versatile storage options.
  • Extra-wide stacking ledge ensures secure and stable stacking.
  • Large label slot allows for clear product identification.
  • Reinforced ribs prevent bins from spreading when fully loaded.
Catalog No. 50-005-29514 Supplier MSC Supplier No. 43383710
This shelf bin is constructed from durable polypropylene and designed for efficient storage. It features a stone color and outer dimensions of 10-7/8 in. L x 4-1/8 in. W x 4 in. H, making it suitable for various organizational needs.
  • Can be hung or stacked for versatile storage options.
  • Extra-wide stacking ledge ensures secure and stable stacking.
  • Large label slot allows for clear product identification.
  • Reinforced ribs prevent bins from spreading when fully loaded.