Learn More
MSC Akro-Mils Shelf Bin, Yellow, Industrial Grade Polymers, 4 in H x 6-5/8 in W x 17-7/8 in D
Catalog No. 5000532206
Encompass
This shelf bin is constructed from durable industrial-grade polymers, providing a neat and attractive storage solution. Its one-piece construction makes it waterproof and resistant to grease and oil. Designed to work with standard wire or steel shelving, this bin can withstand temperatures down to -20° F.
  • Constructed from durable industrial-grade polymers
  • Provides a neat and attractive storage solution
  • Waterproof and unaffected by grease or oil
  • Works with standard wire or steel shelving
  • Works in temperatures down to -20° F
  • One-piece construction
  • Available in 12, 18, and 24-inch depths
  • Available in other colors including Blue, Clear, and Red
  • Buy American Act (BAA) compliant
  • Trade Agreement Act (TAA) compliant
Catalog No. 50-005-32206 Supplier MSC Supplier No. 43185776
This shelf bin is constructed from durable industrial-grade polymers, providing a neat and attractive storage solution. Its one-piece construction makes it waterproof and resistant to grease and oil. Designed to work with standard wire or steel shelving, this bin can withstand temperatures down to -20° F.
  • Constructed from durable industrial-grade polymers
  • Provides a neat and attractive storage solution
  • Waterproof and unaffected by grease or oil
  • Works with standard wire or steel shelving
  • Works in temperatures down to -20° F
  • One-piece construction
  • Available in 12, 18, and 24-inch depths
  • Available in other colors including Blue, Clear, and Red
  • Buy American Act (BAA) compliant
  • Trade Agreement Act (TAA) compliant