Learn More
MSC Akro-Mils Storage Bin, Plastic, Rectangle, Stackable, 60 lb Capacity
Catalog No. 5000511894
Encompass
This plastic storage bin is designed for heavy-duty use in industrial and laboratory settings. It can be stacked for efficient space utilization or hung from louvered racks, panels, rail racks, and carts for high-density storage. Its durable construction makes it corrosion-proof and resistant to weak acids and alkalis.
  • Heavy-duty construction
  • Stackable for efficient storage
  • Hangs from louvered panels and racks
  • Hopper front allows easy access to contents
  • Fits on shelving
  • Corrosion-proof and resistant to weak acids and alkalis
  • 60 lb capacity
  • Length dividers available for organization
Catalog No. 50-005-11894 Supplier MSC Supplier No. 43388107
This plastic storage bin is designed for heavy-duty use in industrial and laboratory settings. It can be stacked for efficient space utilization or hung from louvered racks, panels, rail racks, and carts for high-density storage. Its durable construction makes it corrosion-proof and resistant to weak acids and alkalis.
  • Heavy-duty construction
  • Stackable for efficient storage
  • Hangs from louvered panels and racks
  • Hopper front allows easy access to contents
  • Fits on shelving
  • Corrosion-proof and resistant to weak acids and alkalis
  • 60 lb capacity
  • Length dividers available for organization