Learn More
Enterprise Technology Solutions Akro-Mils Super-Size Storage Bin, Stackable, 18-1/2"x29"x12", BE
SDP
Catalog No. 5000832574
Encompass_Preferred
Control inventories, shorten assembly times and minimize part-handling with this Super-Size AkroBin. Heavy-duty bins securely stack atop each other or sit on shelving. Molded of high-density, industrial-grade polymer, they are unaffected by weak acids and alkalis. Sturdy, one-piece construction is waterproof, rustproof and corrosionproof. Equipped with a large molded-in handle, the bin is easy to carry.
Catalog No. 50-008-32574 Supplier Enterprise Technology Solutions Supplier No. AKM30290B
Control inventories, shorten assembly times and minimize part-handling with this Super-Size AkroBin. Heavy-duty bins securely stack atop each other or sit on shelving. Molded of high-density, industrial-grade polymer, they are unaffected by weak acids and alkalis. Sturdy, one-piece construction is waterproof, rustproof and corrosionproof. Equipped with a large molded-in handle, the bin is easy to carry.