Learn More
Enterprise Technology Solutions Akro-Mils Supply Bin, 5-1/2"x10-7/8"x5-1/4", Clear
SDP
Catalog No. 5000832017
Encompass_Preferred
InSight ultra-clear bin provides attractive, convenient storage. Transparent, autoclavable polycarbonate material provides maximum visibility and easy identification of supplies. Bins can be securely stacked even with lids attached. Strong molded hanger easily hooks onto rails or louvered panels for added storage versatility. Six plastic button feet line the bottom to stabilize the bin on flat or wire shelves. The front of the bin offers a convenient labeling area. Lid is sold separately.
Catalog No. 50-008-32017 Supplier Enterprise Technology Solutions Supplier No. AKM305B1
InSight ultra-clear bin provides attractive, convenient storage. Transparent, autoclavable polycarbonate material provides maximum visibility and easy identification of supplies. Bins can be securely stacked even with lids attached. Strong molded hanger easily hooks onto rails or louvered panels for added storage versatility. Six plastic button feet line the bottom to stabilize the bin on flat or wire shelves. The front of the bin offers a convenient labeling area. Lid is sold separately.