Learn More
Grainger Hang and Stack Bin, 11X16 1/2X18, 1.67 cu ft Capacity, Blue
Catalog No. NC9900873
Encompass_Preferred
AKRO BIN PP 11X16 1/2X18 B,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage. Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF
Catalog No. NC9900873 Supplier Grainger Supplier No. 5YM94
AKRO BIN PP 11X16 1/2X18 B,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage. Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF