Learn More
Grainger F8699 Hang/Stack Bin 10-7/8 x 11 x 5 Blue
Catalog No. NC9865423
Encompass_Preferred
Blue Akro Bin,5x11x10-7/8 in.,resist acids and alkalis; guaranteed against breakage,Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250 deg. F,Reinforced ribs to keep bins from spreading
Catalog No. NC9865423 Supplier Grainger Supplier No. 5W867
Blue Akro Bin,5x11x10-7/8 in.,resist acids and alkalis; guaranteed against breakage,Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250 deg. F,Reinforced ribs to keep bins from spreading