Learn More
Grainger AKROBIN 5X5 1/2X10 7/8 B
Catalog No. NC9785026
Encompass_Preferred
AKRO BIN PP 5X5 1/2X10 7/8 BLUE,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage. Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF
Catalog No. NC9785026 Supplier Grainger Supplier No. 2W778
AKRO BIN PP 5X5 1/2X10 7/8 BLUE,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage. Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF