Learn More
Grainger F8696 Hang/Stack Bin 7x16 1/2x14 3/4 Blue
Catalog No. NC9873475
Encompass_Preferred
AKRO BIN PP 7X16 1/4X14 3/4 BLUE,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage,Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF
Catalog No. NC9873475 Supplier Grainger Supplier No. 2W780
AKRO BIN PP 7X16 1/4X14 3/4 BLUE,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage,Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF