Learn More
Grainger AKROBIN 7X16 1/4X14 3/4 R
Catalog No. NC9674601
Encompass_Preferred
Akrobin, Red, 7 in. x 16- 1/4 in. x 14- 3/4 in., Resist Acids and alkalis, Guaranteed Against Breakage, Heavy-duty Polypropylene Construction is Water, Rust and Corrosion-Proof, and Autoclavable up to 250degreeF, Reinforced Ribs
Catalog No. NC9674601 Supplier Grainger Supplier No. 2W076
Akrobin, Red, 7 in. x 16- 1/4 in. x 14- 3/4 in., Resist Acids and alkalis, Guaranteed Against Breakage, Heavy-duty Polypropylene Construction is Water, Rust and Corrosion-Proof, and Autoclavable up to 250degreeF, Reinforced Ribs