Learn More
Grainger F8697 Hang/Stack Bin 7 x8 1/4 x14 3/4 Red
Catalog No. NC9866587
Encompass_Preferred
AKRO BIN PP 7X8 1/4X14 3/4 RED,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage,Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF
Catalog No. NC9866587 Supplier Grainger Supplier No. 2W075
AKRO BIN PP 7X8 1/4X14 3/4 RED,1/pack,Durable AkroBins resist acids and alkalis and are guaranteed against breakage,Heavy-duty polypropylene construction is water-,rust-,and corrosion-proof,and autoclavable up to 250degreeF