Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Multispecies AKT/ERK Activation InstantOne™ ELISA Kit

Catalog No. 50246233
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-233 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-246-233 Supplier Invitrogen™ Supplier No. 858601411
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 and AKT 1/2/3 in cell lysates. Detection of rat ERK 1/2 and AKT 1/2/3 is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

Extracellular signal-regulated kinase (ERK) is the founding member and a key component of the classical Mitogen-Activated Protein Kinase (MAPK) pathway. ERK1 (p44, MAPK3) and ERK2 (p42, MAPK2) are both activated via the MAPK/ERK pathway, which is downstream of various Receptor Tyrosine Kinases (RTKs) or GPCRs. MEK, a Ser/Thr and Tyr kinase, activates ERK by phosphorylation of its TxY motifs, namely Thr202/Tyr204 and Thr185/Tyr187 of ERK1 and ERK2, respectively. Once activated, ERK phosphorylates a variety of proteins that regulate cellular processes such as cell division, proliferation, survival, differentiation, apoptosis, motility, and metabolism. AKT, which exists as multiple isoforms, is one of the principle kinases activated by phosphoinositide 3-kinase (PI3K). Activation of PI3K results in the generation of phosphatidylinositol (3,4,5)-triphosphate (PIP3). These lipid second messengers bind to the pleckstrin homology domain of AKT to promote its translocation to the plasma membrane for activation via phosphorylation at Thr308 and Ser473 by PDK1 and the mTOR TORC2 complex, respectively. Phosphorylation at both these sites is required for full activation of AKT Ser/Thr kinase activity. AKT phosphorylates over 50 known substrates, including GSK3, AS160, PRAS40, TSC 1, TCS 2, Raf-1, Bad, the FOXO family of transcription factors, and PFK2. Due to its established critical role in numerous biological processes, AKT is a frequent target for oncology-related drug development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27361, P28482, P31749, P31751, Q9Y243
Content And Storage • User Manual
• phospho-AKT 1/2/3 (Ser437) Capture Antibody Reagent, 1 x 1.5 mL
• phospho-AKT 1/2/3 (Ser437) Detection Antibody Reagent, 1 x 1.5 mL
• phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Capture Antibody Reagent, 1 x 1.5 mL
• phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Detection Antibody Reagent, 1 x 1.5 mL
• InstantOne ELISA Assay Plate, 96 well (sealed)
• Cell Lysis Buffer (5X), 10 mL
• Enhancer Solution, 1 mL
• Wash Buffer (10X), 15 mL
• Detection Reagent (TMB Substrate), 12 mL
• Stop Solution, 12 mL
• Positive Control Cell Lysate (lyophilized), 0.25 mL
• Plate seal, 1 ea
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ERK; pan AKTERK; pan AKT; AKT; CWS6; PKB; PKB-ALPHA; PRKBA; RAC; RAC-ALPHA;ERK; ERK-2; ERK2; ERT1; MAPK2; P42MAPK; PRKM1; PRKM2; p38; p40; p41; p41mapk; p42-MAPK;ERK-1; ERK1; ERT2; HS44KDAP; HUMKER1A; P44ERK1; P44MAPK; PRKM3; p44-ERK1; p44-MAPK
Gene ID (Entrez) 10000, 207, 208, 5594, 5595
Gene Symbol AKT1, AKT2, AKT3, MAPK1, MAPK3
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOneTM Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2-8°C
Synonym PKB, RAC-Pk.-alpha / p44/p42, MAPK3/MAPK2
Target AKT Pan/Erk1/2
Test Time 1 hr 20 min
Total Assay Time 1 hr 10 min
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.