Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AKT-IN-23 | 1226801-23-5 | 99.0% | 517.52 g/mol | C25H27F4N7O | 10 MG
SDP

Catalog No. 5000288779
Encompass_Preferred

AKT-IN-23 is a small-molecule AKT inhibitor provided as a white to off-white solid for preclinical and cancer research applications. It is formulated for use in biochemical and cellular assays that probe AKT signaling pathways.

  • High purity: 98.96%.
  • Molecular weight: 517.52 g/mol.
  • Chemical formula: C25H27F4N7O.
  • Soluble in DMSO at approximately 25 mg/mL.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities (mg scale).

Catalog No. 50-002-88779 Supplier Medchemexpress LLC Supplier No. HY16170410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AKT-IN-23 is a small-molecule AKT inhibitor provided as a white to off-white solid for preclinical and cancer research applications. It is formulated for use in biochemical and cellular assays that probe AKT signaling pathways.

  • High purity: 98.96%.
  • Molecular weight: 517.52 g/mol.
  • Chemical formula: C25H27F4N7O.
  • Soluble in DMSO at approximately 25 mg/mL.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities (mg scale).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.