Learn More
Medchemexpress LLC AKT-IN-6 | 1430056-54-4 | 99.1% | 389.43 | 1 ML
SDP
Catalog No. 5000402039
Encompass_Preferred
AKT-IN-6 is a potent Akt inhibitor that targets Akt1, Akt2, and Akt3 with IC50s below 500 nM. Akt is a central node in cell signaling, and its aberrant activation is linked to various complex diseases, including type-2 diabetes and cancer.
  • Potent Akt inhibitor
  • Targets Akt1, Akt2, and Akt3
  • Suitable for research applications
Catalog No. 50-004-02039 Supplier Medchemexpress LLC Supplier No. HY1998210MM/1ML
AKT-IN-6 is a potent Akt inhibitor that targets Akt1, Akt2, and Akt3 with IC50s below 500 nM. Akt is a central node in cell signaling, and its aberrant activation is linked to various complex diseases, including type-2 diabetes and cancer.
  • Potent Akt inhibitor
  • Targets Akt1, Akt2, and Akt3
  • Suitable for research applications