Learn More
Medchemexpress LLC Akt-in-6 | 1430056-54-4 | 99.1% | 389.43 | 5 MG
SDP
Catalog No. 5000403980
Encompass_Preferred
AKT-IN-6 is a potent Akt inhibitor that inhibits Akt1, Akt2, and Akt3 with IC50s less than 500 nM.
  • Potent Akt inhibitor.
  • Inhibits Akt1, Akt2, and Akt3.
  • Available in solid form (yellow to brown color).
  • Soluble in DMSO (125 mg/mL).
  • Multiple package sizes are available (5 mg to 100 mg).
  • Associated with various related screening libraries, small molecules, and proteins.
Catalog No. 50-004-03980 Supplier Medchemexpress LLC Supplier No. HY199825MG
AKT-IN-6 is a potent Akt inhibitor that inhibits Akt1, Akt2, and Akt3 with IC50s less than 500 nM.
  • Potent Akt inhibitor.
  • Inhibits Akt1, Akt2, and Akt3.
  • Available in solid form (yellow to brown color).
  • Soluble in DMSO (125 mg/mL).
  • Multiple package sizes are available (5 mg to 100 mg).
  • Associated with various related screening libraries, small molecules, and proteins.