Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs AKT INHIBITOR (FPA-124) 5 mg
SDP

Catalog No. 50720006
Encompass

FPA-124 is a cell-permeable inhibitor of Akt (IC50 = 100 nM), binding to the PH and kinase domains. It inhibits cell proliferation in Colo357, BxPC3, BT20, and PC3 cancer cell lines with IC50 values ranging from 7-55 nM. FPA-124 inhibited tumor growth in mice with negligible toxicity.nn


nn

Solubility

nDMSO: 5 mg/mLn

Storage

nStore dry between 2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) V. Barve, et al. "Synthesis, Molecular Characterization, and Biological Activity of Novel Synthetic This item is non returnable

Catalog No. 50-720-006 Supplier Echelon Biosciences Research Labs Supplier No. B01015MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FPA-124 is a cell-permeable inhibitor of Akt (IC50 = 100 nM), binding to the PH and kinase domains. It inhibits cell proliferation in Colo357, BxPC3, BT20, and PC3 cancer cell lines with IC50 values ranging from 7-55 nM. FPA-124 inhibited tumor growth in mice with negligible toxicity.nn


nn

Solubility

nDMSO: 5 mg/mLn

Storage

nStore dry between 2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) V. Barve, et al. "Synthesis, Molecular Characterization, and Biological Activity of Novel Synthetic This item is non returnable

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.