Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs AKT INHIBITOR (KP372-1) 5 mg
SDP

Supplier:  Echelon Biosciences Research Labs B01025MG

Encompass

KP372-1 directly inhibits the kinase activities of Akt, PDK1 and Flt3 (Fms-like tyrosine kinase 3), which are aberrantly activated in acute myelogenous leukemia (AML). It is a cell permeant inhibitor that decreases the colony-forming ability of AML cells (IC50 < 200 nM) with minimal cytoxic effects on normal hematopoietic progenitor cells. KP372-1 has also been shown to suppress cell proliferation and induce apoptosis in thyroid cancer cells and gliobastoma. Provided as a 1:1 mixture of regioisomers.nn


nn

Solubility

nWater: insoluble, DMSO: 5 mg/mLn

Storage

n2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) M. This item is non returnable

Catalog No. 50-720-007


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.