Learn More
Enzo Life Sciences Akt inhibitor VIII (5mg). CAS: 612847-09-3
SDP
Catalog No. 502013414
Encompass_Preferred
A cell-permeable compound that potently, selectively and reversibly inhibits Akt1/Akt2/Akt3 activity. Alternative name: Akti-1/2, Isozyme-selective Akt inhibitor VIII, 1,3-Dihydro-1-(1-((4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenyl)methyl)-4-piperidinyl)-2H-benzimidazol-2-one, Imidazoquinoxaline 16h. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.
Catalog No. 50-201-3414 Supplier Enzo Life Sciences Supplier No. ENZCHM1250005
A cell-permeable compound that potently, selectively and reversibly inhibits Akt1/Akt2/Akt3 activity. Alternative name: Akti-1/2, Isozyme-selective Akt inhibitor VIII, 1,3-Dihydro-1-(1-((4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenyl)methyl)-4-piperidinyl)-2H-benzimidazol-2-one, Imidazoquinoxaline 16h. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.