Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AKT inhibitor VIII | 612847-09-3 | MFCD23843802 | 99.0% | 551.64 g/mol | C34H29N7O | 100 MG
SDP

Catalog No. 5000247829
Encompass_Preferred

AKT inhibitor VIII is a cell-permeable, reversible allosteric inhibitor of Akt kinases used as a research reagent to probe PI3K/Akt signaling pathways. It is supplied as a light yellow solid for laboratory use.

  • Allosteric, reversible Akt inhibitor.
  • Isoform-selective activity with greater potency for Akt1 and Akt2.
  • Reported IC50 values: Akt1 = 58 nM, Akt2 = 210 nM.
  • Molecular formula C34H29N7O; molecular weight 551.64 g/mol.
  • High reported purity (~99%).
  • Supplied as a light yellow solid with recommended storage for powder at -20°C or 4°C depending on duration.

Catalog No. 50-002-47829 Supplier Medchemexpress LLC Supplier No. HY10355100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AKT inhibitor VIII is a cell-permeable, reversible allosteric inhibitor of Akt kinases used as a research reagent to probe PI3K/Akt signaling pathways. It is supplied as a light yellow solid for laboratory use.

  • Allosteric, reversible Akt inhibitor.
  • Isoform-selective activity with greater potency for Akt1 and Akt2.
  • Reported IC50 values: Akt1 = 58 nM, Akt2 = 210 nM.
  • Molecular formula C34H29N7O; molecular weight 551.64 g/mol.
  • High reported purity (~99%).
  • Supplied as a light yellow solid with recommended storage for powder at -20°C or 4°C depending on duration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.