Learn More
STEMCELL Technologies AKT Inhibitor VIII, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72944
Encompass
AKT Inhibitor VIII is a cell-permeable, allosteric inhibitor of all three forms of the kinase AKT (AKT1, AKT2, and AKT3) with IC₅₀ values of 58, 210, and 2,200 nM, respectively (Lindsley et al.; Calleja et al.). It displays good selectivity against a panel of 70 other kinases with micromolar inhibition against some kinases, such as calcium/calmodulin-dependent protein kinase 1 and smooth muscle myosin light-chain kinase (Logie et al.).   CANCER RESEARCH· Sensitizes prostate tumor and cervical carcinoma cells to apoptotic stimuli (DeFeo-Jones et al.).· Blocks mitosis and inhibits migration of HeLa cells (Jo et al.). METABOLISM· Reduces insulin-dependent gene repression in liver cells leading to reduced insulin sensitivity (Logie et al.).
Catalog No. 50-197-6004
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov