Learn More
Cayman Chemical Akt Inhibitor VIII, Purity: ≥98%, CAS: 612847-09-3, 1 mg
SDP
Catalog No. 502969999
Encompass_Preferred
A potent, allosteric inhibitor of Akt1 and Akt2 (IC50 = 58 and 210 nM, respectively) that less effectively blocks Akt3 activity (IC50 = 2.2 µM); cell permeable, blocking insulin regulation of FOXO1 activity at 1 µM in rat hepatoma cells
Catalog No. 50-296-9999 Supplier Cayman Chemical Supplier No. 148701 MG
A potent, allosteric inhibitor of Akt1 and Akt2 (IC50 = 58 and 210 nM, respectively) that less effectively blocks Akt3 activity (IC50 = 2.2 µM); cell permeable, blocking insulin regulation of FOXO1 activity at 1 µM in rat hepatoma cells