Learn More
Cayman Chemical Akt InhIbItr VIII 500ug
SDP
Catalog No. 502970823
Encompass_Preferred
A potent, allosteric inhibitor of Akt1 and Akt2 (IC50 = 58 and 210 nM, respectively) that less effectively blocks Akt3 activity (IC50 = 2.2 µM); cell permeable, blocking insulin regulation of FOXO1 activity at 1 µM in rat hepatoma cells
Catalog No. 50-297-0823 Supplier Cayman Chemical Supplier No. 14870500 UG
Add to Cart
Add to Cart
A potent, allosteric inhibitor of Akt1 and Akt2 (IC50 = 58 and 210 nM, respectively) that less effectively blocks Akt3 activity (IC50 = 2.2 µM); cell permeable, blocking insulin regulation of FOXO1 activity at 1 µM in rat hepatoma cells