Learn More
Cayman Chemical Akt InhIbItr XI 1mg
SDP
Catalog No. 502967343
Encompass_Preferred
A cell-permeable, copper-containing 3-formylchromone derivative that inhibits Akt in an array of cancer cells (IC50s = 10-34 µM); also causes NF-κB inactivation in an orthotopic pancreatic tumor model using COLO 357 cells
Catalog No. 50-296-7343 Supplier Cayman Chemical Supplier No. 186041 MG
A cell-permeable, copper-containing 3-formylchromone derivative that inhibits Akt in an array of cancer cells (IC50s = 10-34 µM); also causes NF-κB inactivation in an orthotopic pancreatic tumor model using COLO 357 cells