Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Akt kinase inhibitor hydrochloride | 3026697-00-4 | MFCD34676556 | 99.4% | 393.83 g/mol | C16H20ClN7O3 | 10 MG
SDP

Catalog No. 5000246727
Encompass_Preferred

AKT Kinase Inhibitor (hydrochloride) is a small-molecule Akt pathway inhibitor supplied for research use. Provided as the hydrochloride salt, it is used in biochemical and cellular assays to inhibit Akt kinase activity and study signaling and oncology-related pathways.

  • High reported purity (99.4%).
  • Molecular weight 393.83 g/mol.
  • Soluble in DMSO and water (DMSO 3.33 mg/mL; H2O 20 mg/mL).
  • Supplied in milligram-scale quantities suitable for assay development and in vitro studies.
  • Refrigerated storage recommended per supplier guidance.

Catalog No. 50-002-46727 Supplier Medchemexpress LLC Supplier No. HY10249D10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AKT Kinase Inhibitor (hydrochloride) is a small-molecule Akt pathway inhibitor supplied for research use. Provided as the hydrochloride salt, it is used in biochemical and cellular assays to inhibit Akt kinase activity and study signaling and oncology-related pathways.

  • High reported purity (99.4%).
  • Molecular weight 393.83 g/mol.
  • Soluble in DMSO and water (DMSO 3.33 mg/mL; H2O 20 mg/mL).
  • Supplied in milligram-scale quantities suitable for assay development and in vitro studies.
  • Refrigerated storage recommended per supplier guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.