Learn More
Enzo Life Sciences Akt polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPPK004F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Akt. The sequence is completely conserved in rat and bovine. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7290