Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ AKT Phospho-Specific Array

Catalog No. 89933040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit (2 Samples)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-040 1 Kit (2 Samples)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-040 Supplier Abnova™ Supplier No. AA0012
Add to Cart
Add to Cart

AKT Phospho-Specific Array includes 137 highly specific and well-characterized phosphorylation antibodies in the AKT pathway.

  • This product must be accompanied with Antibody Array Detection Kit
  • The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
  • Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum

Specifications

Content And Storage 4°C
Detection Method Fluorometric
Quantity 1 Kit (2 Samples)
For Use With (Application) Profiling
Product Type Array/Library
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.