Learn More
CPC Scientific H-Ala-Arg-Lys-Arg-Glu-Arg-Thr-Tyr-Ser(PO3H2)-Phe-Gly-His-His-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST078A
Encompass
The native peptide ARKRERTYSFGHHA (29944-1) is a synthetic substrate for AKT/PKB/Rac-protein kinases. Phophorylation is at the Ser site (Km = 3.9 M). It also competitively inhibits histone H2B phosphorylation (Ki = 12 M) by AKT.SEQUENCE: H-Ala-Arg-Lys-Arg-Glu-Arg-Thr-Tyr-Ser(PO3H2)-Phe-Gly-His-His-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: ARKRERTY-pS-FGHHAMOLECULAR FORMULA: MOLECULAR WEIGHT: 1795.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:Obata, T. et al. J. Biol. Chem. 275, 36108 (2000) Chan, TO. et al. Ann. Rev. Biochem. 68, 965 (1999).SKU(s): HIST-078A
Catalog No. 50-210-9520