Learn More
Sino Biological Peptide Substrates: AKT (PKB) Substrate
Catalog No. 502569867
Encompass_Preferred
The AKT (PKB) Substrate peptide sequence (CKRPRAASFAE) is based on the N-terminus of GSK3.
Catalog No. 50-256-9867 Supplier Sino Biological Supplier No. A0558
The AKT (PKB) Substrate peptide sequence (CKRPRAASFAE) is based on the N-terminus of GSK3.