Learn More
Enzo Life Sciences Akt (pSer473) monoclonal antibody (11E6) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804401C100
Encompass_Preferred
Clone: 11E6. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to C-terminal (PQFpSYSA) of human Akt (protein kinase B; PKB) phosphorylated at Ser473 conjugated to KLH. Species reactivity: Human, Mouse, Rat, Dog. Specificity: Recognizes Ser472/473/474-phosphorylated Akt (see technical note). Applications: ELISA, Flow Cytometry, IHC, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9901