Learn More
Enzo Life Sciences Akt (pSer473) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905773100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Akt1 phosphorylated at Ser473. UniProt ID: P31749. Source: Purified from rabbit serum. Species reactivity: Mouse. Creossreactivity: Weakly cross-reacts with human, rat, bovine and chicken Akt. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7047