Learn More
Enzo Life Sciences Akt Recombinant monoclonal antibody (RM316) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS6740100
Encompass_Preferred
Clone: RM316. Isotype: IgG. Immunogen: Synthetic peptide corresponding to the PH domain of human AKT1/2/3. Source: Purified from animal origin-free supernatant. Species reactivity: Human. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In 1x PBS containing 50% glycerol, 1% BSA and 0.09% sodium azide.
Catalog No. 50-201-3202