Learn More
Medchemexpress LLC HY-50862 5mg Medchemexpress, Akt1 and Akt2-IN-1 CAS:893422-47-4 Purity:>98%
SDP
Catalog No. 502082257
Encompass_Preferred
Medchemexpress, HY-50862 5mg Akt1 and Akt2-IN-1 CAS:893422-47-4 Akt1 and Akt2-IN-1 is an allosteric inhibitor of Akt1 (IC50=3.5 nM) and Akt2 (IC50=42 nM), with potent and balanced activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2257 Supplier Medchemexpress LLC Supplier No. HY508625MG
Add to Cart
Add to Cart
Medchemexpress, HY-50862 5mg Akt1 and Akt2-IN-1 CAS:893422-47-4 Akt1 and Akt2-IN-1 is an allosteric inhibitor of Akt1 (IC50=3.5 nM) and Akt2 (IC50=42 nM), with potent and balanced activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.